missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Epsin 1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA5122012
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Epsin 1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a member of the epsin protein family. The encoded protein binds clathrin and is involved in the endocytosis of clathrin-coated vesicles. Loss of function of this gene is associated with reduced tumor growth and progression in certain cancer types.
Spécification
| Epsin 1 | |
| Polyclonal | |
| Unconjugated | |
| EPN1 | |
| AA269831; EH domain-binding mitotic phosphoprotein; Epn1; EPS-15-interacting protein 1; Epsin 1; epsin-1; hypothetical protein LOC508199; Ibp1; Intersectin-EH-binding protein 1; LOW QUALITY PROTEIN: epsin-1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 13854, 29924 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q80VP1, Q9Y6I3 | |
| EPN1 | |
| Recombinant protein Epsin 1. The antigen corresponds to amino acid range 279-437 of the target protein. | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu