Learn More
ErbB3/Her3 Antibody (S05-3F7), Novus Biologicals™
Rabbit Monoclonal Antibody
Marque: Novus Biologicals NBP3-19862-50ul
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
ErbB3/Her3 Monoclonal antibody specifically detects ErbB3/Her3 in Human, Mouse samples. It is validated for Western Blot
Spécification
| ErbB3/Her3 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| S05-3F7 | |
| Unconjugated | |
| c-erbB3, EC 2.7.10, EC 2.7.10.1, ErbB-3, erbB3-S, HER3c-erbB-3, LCCS2, lethal congenital contracture syndrome 2, MDA-BF-1, MGC88033, p180-ErbB3, p45-sErbB3, p85-sErbB3, Proto-oncogene-like protein c-ErbB-3, receptor tyrosine-protein kinase erbB-3, Tyrosine kinase-type cell surface receptor HER3, v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian) | |
| A synthetic peptide of human ErbB3/Her3 (Uniprot # P21860) | |
| 50 μg | |
| Cancer, Phospho Specific, Protein Kinase, Tumor Suppressors | |
| 2065 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |