Learn More
Estrogen Receptor, alpha (Marker of Estrogen Dependence) Mouse, Clone: ESR1/1935, enQuireBio™
Mouse Monoclonal Antibody
Marque: EnquireBio 2099-MSM15-P0
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Spécification
| Estrogen Receptor, alpha (Marker of Estrogen Dependence) | |
| Nucleus | |
| ESR1/1935 | |
| Unconjugated | |
| Mouse | |
| ∽67kDa | |
| Protein A/G purified from Bioreactor concentrate. | |
| RUO | |
| 2099 | |
| IgG2a κ |
| Western Blot, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant full-length human Estrogen Receptor alpha protein | |
| MCF-7 Cells. Breast or Endometrial Carcinoma. | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |