missing translation for 'onlineSavingsMsg'
Learn More

Exosome component 1 Antibody [DyLight 650], Novus Biologicals Biologicals™

Code produit 30497928 Tous les produits Bio Techne Produits
Modifier l'affichage
Click to view available options
Quantité:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité
30497928 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 30497928 Fournisseur Novus Biologicals Code fournisseur NBP335122C
 Article restreint
Expédition estimée: 15-09-2026
Ajouter au panier
Ajouter au panier
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Polyclonal Antibody

Exosome component 1 Polyclonal antibody specifically detects Exosome component 1 in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigène Exosome component 1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugué DyLight 650
Formule 50mM Sodium Borate
Alias de gène CGI-108,3'-5' exoribonuclease CSL4 homolog, CSL4 exosomal core protein homolog, Csl4p, CSL4p13, exosomal core protein CSL4, exosome component 1hCsl4p, homolog of yeast exosomal core protein CSL4, SKI4, Ski4p
Espèces hôtes Rabbit
Immunogène Recombinant fusion protein containing a sequence corresponding to amino acids 1-195 of human Exosome component 1 (NP_057130.1).,, Sequence:, MAPPVRYCIPGERLCNLEEGSPGSGTYTRHGYIFSSLAGCLMKSSENGALPVVSVVRETESQLLPDVGAIVTCKVSSINSRFAKVHILYVGSMPLKNSFRGTIRKEDVRATEKDKVEIYKSFRPGDIVLAKVISLGDAQSNYLLTTAENELGVVVAHSESGIQMVPISWCEMQCPKTHTKEFRKVARVQPEFLQT
Méthode de purification Affinity purified
Quantité 0.1 mL
État réglementaire RUO
Disciplines de recherche Immunology
Primaire ou secondaire Primary
Identification génétique (Entrez) 51013
Espèces cibles Human
Contenu et stockage Store at 4°C in the dark.
Type de produit Antibody
Forme Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.