missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EZH2/KMT6 Antibody (CL11915), Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP3-21204-100ul
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
EZH2/KMT6 Monoclonal antibody specifically detects EZH2/KMT6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Spécification
| EZH2/KMT6 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| CL11915 | |
| Western Blot 1 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 2-10 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| EC 2.1.1, EC 2.1.1.43, enhancer of zeste (Drosophila) homolog 2, enhancer of zeste 2, Enhancer of zeste homolog 2, enhancer of zeste homolog 2 (Drosophila), ENX1, ENX-1KMT6EZH1KMT6A, histone-lysine N-methyltransferase EZH2, Lysine N-methyltransferase 6, MGC9169 | |
| This antibody was developed against a Synthetic Peptide of Q15910. Actual immunogen sequence is proprietary. | |
| 100 μg | |
| Breast Cancer, Cancer, Chromatin Research, Phospho Specific, Prostate Cancer | |
| 2146 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu