missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ F4/80 Monoclonal Antibody (BM8), Alexa Fluor™ 647

Rat Monoclonal Antibody
Marque: Invitrogen™ MF48021
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
F4/80 Monoclonal Antibody for Flow
F4/80 antigen is a 160 kDa cell surface glycoprotein that is a member of the EGF TM7 family of proteins and shares 68% overall amino acid identity with human EGF module containing mucin like hormone receptor 1 (EMR1). Expression of F4/80 is heterogeneous and is reported to vary during macrophage maturation and activation. The F4/80 antigen is expressed on a wide range of mature tissue macrophages including Kupffer cells, Langerhans, microglia, macrophages located in the gut lamina propria, peritoneal cavity, lung, thymus, bone marrow stroma and macrophages in the red pulp of the spleen. F4/80 expression has also been reported on a subpopulation of dendritic cells but is absent from macrophages located in T cell areas of the spleen and lymph node. The ligands and biological functions of the F4/80 antigen have not yet been determined but recent studies suggest a role for F4/80 in the generation of efferent CD8+ve regulatory T cells.
Spécification
| F4/80 | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 4mg/mL BSA and 0.1% sodium azide | |
| Q61549 | |
| ADGRE1 | |
| mouse F4/80 which is a macrophage-restricted cell surface glycoprotein. | |
| 1 mL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Flow Cytometry | |
| BM8 | |
| Alexa Fluor 647 | |
| ADGRE1 | |
| ADGRE1; Adhesion G protein-coupled receptor E1; Adhesion G protein-coupled receptor E2; cell surface glycoprotein EMR1; cell surface glycoprotein F4/80; DD7A5-7; EGF-like module containing, mucin-like, hormone receptor-like 1; EGF-like module containing, mucin-like, hormone receptor-like sequence 1; EGF-like module receptor 1; EGF-like module-containing mucin-like hormone receptor-like 1; EGF-TM7; Emr1; EMR1 hormone receptor; F4/80; Gpf480; Ly71; lymphocyte antigen 71; TM7LN3 | |
| Rat | |
| Purified | |
| RUO | |
| 13733 | |
| 4°C | |
| Liquid | |
| Responsibly packaged |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu