missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
FAF1 Monoclonal specifically detects FAF1 in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Protein Array.
Spécification
Specifications
| Antigène | FAF1 |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Peptide Array |
| Classification | Monoclonal |
| Clone | CPTC-FAF1-2 |
| Concentration | 0.2 mg/mL |
| Conjugué | Unconjugated |
| Dilution | Western Blot 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array |
| Formule | 10 mM PBS with 0.05% BSA |
| Alias de gène | CGI-03, Fas (TNFRSF6) associated factor 1, HFAF1s, TNFRSF6-associated factor 1, UBX domain protein 3A, UBX domain-containing protein 12, UBX domain-containing protein 3A, UBXD12FLJ37524, UBXN3AhFAF1FAS-associated factor 1 |
| Espèces hôtes | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?