missing translation for 'onlineSavingsMsg'
Learn More

FAM193A Antibody, Novus Biologicals™

Product Code. p-200046067 Shop All Bio Techne Products
Change view
Click to view available options
Quantité:
0.1 mL
25 μL
Unit Size:
0.10mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité unitSize
18493782 25 μL 25µL
18385571 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18493782 Supplier Novus Biologicals Supplier No. NBP19116525ul

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FAM193A Polyclonal specifically detects FAM193A in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigène FAM193A
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugué Unconjugated
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500
Formule PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Alias de gène family with sequence similarity 193, member A
Symboles de gène(s) FAM193A
Espèces hôtes Rabbit
Immunogène This antibody was developed against Recombinant Protein corresponding to amino acids:RSISTFLGTLENEHLKKFQVTWELHNKHLFENLVFSEPLLQSNLPALVSQIRLGTTTHDTCSEDTYSTLLQRYQRSEEELRRVAEEWLECQKRIDAYVDEQM
Méthode de purification Affinity Purified
Quantité 25 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 8603
Spécificité du test Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Espèces cibles Human
Contenu et stockage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.