missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FBXW7/Cdc4 Antibody (FBOX 3a9/1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP2-50403
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
FBXW7/Cdc4 Monoclonal antibody specifically detects FBXW7/Cdc4 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, CyTOF, KnockDown
Spécification
| FBXW7/Cdc4 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, CyTOF-ready, Knockdown Validated | |
| AGOFBXO30, Archipelago homolog, archipelago, Drosophila, homolog of, CDC4FLJ16457, F-box and WD repeat domain containing 7, F-box and WD-40 domain protein 7 (archipelago homolog, Drosophila), F-box and WD-40 domain-containing protein 7, F-box protein FBW7, F-box protein FBX30, F-box protein SEL-10, F-box/WD repeat-containing protein 7, FBW7hCdc4, FBX30FLJ11071, FBXW6, hAgo, homolog of C elegans sel-10, SEL-10DKFZp686F23254, SEL10FBW6 | |
| Synthetic peptide of human sequence | |
| 0.1 mL | |
| Cancer | |
| 55294 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| FBOX 3a9/1 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu