missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ FBXW7/Cdc4 Antibody [PE], Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals™ NB100-88139PE
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
FBXW7/Cdc4 Polyclonal antibody specifically detects FBXW7/Cdc4 in Human samples. It is validated for Immunohistochemistry,Immunoprecipitation,Immunohistochemistry (Paraffin)
Spécification
| FBXW7/Cdc4 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| PE | |
| AGOFBXO30, Archipelago homolog, archipelago, Drosophila, homolog of, CDC4FLJ16457, F-box and WD repeat domain containing 7, F-box and WD-40 domain protein 7 (archipelago homolog, Drosophila), F-box and WD-40 domain-containing protein 7, F-box protein FBW7, F-box protein FBX30, F-box protein SEL-10, F-box/WD repeat-containing protein 7, FBW7hCdc4, FBX30FLJ11071, FBXW6, hAgo, homolog of C elegans sel-10, SEL-10DKFZp686F23254, SEL10FBW6 | |
| The immunogen recognized by this antibody maps to a region between residue 50 and 100 of human F-box and WD-40 domain protein 7 using the numbering given in entry NP_361014.1 (GeneID 55294). | |
| 0.1 mL | |
| Cancer | |
| 55294 | |
| Store at 4°C in the dark. | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu