Learn More
FCRN/FCGRT Antibody, Novus Biologicals™
Code produit 18739893 Tous les produits Bio Techne Produits
Change view
Click to view available options
Conditionnement:
0.10mL
25µL
Quantité:
25 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit unitSize Quantité
18739893 0.10mL -
18441971 25µL 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 18739893 Fournisseur Novus Biologicals Code fournisseur NBP189128
Rabbit Polyclonal Antibody
FCRN/FCGRT Polyclonal specifically detects FCRN/FCGRT in Human, Cynomolgus Monkey samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated.
TRUSTED_SUSTAINABILITY
Spécification
Antigène FCRN/FCGRT
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
For Research Use Only