Learn More
Fibronectin Antibody (1E1), Novus Biologicals™
Code produit 30043724 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
50 μL
100 μL
Conditionnement:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30043724 50 μL 50µL
30043534 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 30043724 Fournisseur Novus Biologicals Code fournisseur NBP32657750ul
Rabbit Monoclonal Antibody
Fibronectin Monoclonal antibody specifically detects Fibronectin in Human samples. It is validated for ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Spécification
Antigène Fibronectin
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 1E1
Conjugué Unconjugated
Dilution ELISA, Immunohistochemistry 1:50-1:200
Formule PBS, pH 7.4, 150mM NaCl and 50% glycerol
Alias de gène CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF
Espèces hôtes Rabbit
Immunogène A synthesized peptide derived from Human Fibronectin [UniProt P02751]
Méthode de purification Affinity purified
Quantité 50 μL
État réglementaire RUO
Disciplines de recherche Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primaire ou secondaire Primary
Identification génétique (Entrez) 2335
Espèces cibles Human
Contenu et stockage Store at -20 to -70C. Avoid freeze-thaw cycles.
Forme Purified
Isotype IgG