Learn More
Fibronectin Antibody (A17), DyLight 594, Novus Biologicals™
Code produit 18498936 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
0.1 mL
Conditionnement:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18498936 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 18498936 Fournisseur Novus Biologicals Code fournisseur NBP223524DL594
Mouse Monoclonal Antibody
Fibronectin Monoclonal antibody specifically detects Fibronectin in Human, Bovine, Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY
Spécification
Antigène Fibronectin
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone A17
Conjugué DyLight 594
Formule 50mM Sodium Borate
Alias de gène CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF
Espèces hôtes Mouse
Immunogène Lysed bovine corneal endothelial cells and extracellular matrix
Méthode de purification Protein A or G purified
Quantité 0.1 mL
État réglementaire RUO
Disciplines de recherche Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primaire ou secondaire Primary
Identification génétique (Entrez) 2335
Espèces cibles Human, Bovine, Chicken
Contenu et stockage Store at 4°C in the dark.
Forme Purified
Isotype IgG1