Learn More
ACROBiosystems FITC-Labeled Human DLL3 Protein, Fc Tag
Marque:  ACROBiosystems DL3-HF253-200ug
Informations supplémentaires : Poids : 0.00010kg
 Afficher plus de versions de ce produit
Code produit. 17256190
  • 2486.22€ / 200µg
Expédition estimée: 14-10-2022
pour voir le stock disponible 
Description
Description
Delta-like protein 3 (DLL3) is a transmembrane protein that belongs to the Delta/Serrate/Lag-2 (DSL) family of Notch ligands.May be required to divert neurons along a specific differentiation pathway. Plays a role in the formation of somite boundaries during segmentation of the paraxial mesoderm. DLL3 protein is expressed on the surface of tumor cells but not in normal adult tissues.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 75.0 kDa.
Spécification
Spécification
NP_058637.1
75.0 kDa
FITC-Labeled Human DLL3, Fc Tag (DL3-HF253) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Leu 492 (Accession # Q9NYJ7-1).
-20°C, avoid light
DLL3
Fc Tag
Fluorescent Labeled Protein
Powder
200 μg
Ala 27 - Leu 492
Delta3,delta-like 3 (Drosophila),delta-like protein 3,DLL3,Pudgy,SCDO1,SCDO1delta3
FITC Labeled Protein
HEK293