missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLIP Antibody (NF6), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP2-80081FR
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
FLIP Monoclonal antibody specifically detects FLIP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Spécification
| FLIP | |
| Monoclonal | |
| DyLight 680 | |
| CASHFADD-like antiapoptotic molecule 1, CASP8 and FADD-like apoptosis regulator, CASP8AP1, Caspase homolog, Caspase-eight-related protein, Caspase-like apoptosis regulatory protein, caspase-related inducer of apoptosis, CASPER, CasperFLAME1, Cellular FLICE-like inhibitory protein, c-FLIPR, c-FLIPS, CLARPMACH-related inducer of toxicity, FADD-like apoptotic molecule, FLAME, FLAME-1, I-FLICEMRITc-FLIPFLIPc-FLIPL, Inhibitor of FLICE, Usurpin, usurpin beta | |
| Recombinant human FLIP (aa 1-480). | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Inhibitors Activators and Regulators, Signal Transduction, Tumor Suppressors | |
| 8837 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| NF6 | |
| 50mM Sodium Borate | |
| Mouse | |
| >95%, by SDS-PAGE | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu