Learn More
Invitrogen™ Flotillin 1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA517127
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
It is not recommended to aliquot this antibody.
Caveolae are small domains on the inner cell membrane involved in vesicular trafficking and signal transduction. FLOT1 encodes a caveolae-associated, integral membrane protein. The function of flotillin 1 has not been determined.
Spécification
| Flotillin 1 | |
| Polyclonal | |
| Unconjugated | |
| FLOT1 | |
| DADB-118P11.2; Flot1; flotillin 1; Flotillin-1; I79_013152; integral membrane component of caveolae; OTTHUMP00000164855; Rareg; Reg2; REG-2; reggie-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10211, 14251, 64665 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 63 μg/mL | |
| 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
| O08917, O75955, Q9Z1E1 | |
| FLOT1 | |
| Synthetic peptide corresponding residue surrounding Ile368 of human flotillin-1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Monkey, Rat | |
| Antibody | |
| IgG |