missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ FPGT (Human) Recombinant Protein

Human FPGT full-length ORF ( NP_003829.2, 1 a.a. - 594 a.a.) recombinant protein with GST-tag at N-terminal.

Marque:  Abnova™ H00008790-P01.25ug

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16152902

  • 508.00€ / 25µg
Expédition estimée: 20-06-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

  • Sequence: MAAARDPPEVSLREATQRKLRRFSELRGKLVARGEFWDIVAITAADEKQELAYNQQLSEKLKRKELPLGVQYHVFVDPAGAKIGNGGSTLCALQCLEKLYGDKWNSFTILLIHSGGYSQRLPNASALGKIFTALPLGNPIYQMLELKLAMYIDFPLNMNPGILVTCADDIELYSIGEFEFIRFDKPGFTALAHPSSLTIGTTHGVFVLDPFDDLKHRDLEYRSCHRFLHKPSIEKMYQFNAVCRPGNFCQQDFAGGDIADLKLDSDYVYTDSLFYMDHKSAKMLLAFYEKIGTLSCEIDAYGDFLQALGPGATVEYTRNTSNVIKEESELVEMRQRIFHLLKGTSLNVVVLNNSKFYHIGTTEEYLFYFTSDNSLKSELGLQSITFSIFPDIPECSGKTSCIIQSILDSRCSVAPGSVVEYSRLGPDVSVGENCIISGSYILTKAALPAHSFVCSLSLKMNRCLKYATMAFGVQDNLKKSVKTLSDIKLLQFFGVCFLSCLDVWNLKVTEELFSGNKTCLSLWTARIFPVCSSLSDSVITSLKMLNAVKNKSAFSLNSYKLLSIEEMLIYKDVEDMITYREQIFLEISLKSSLM
Spécification

Spécification

NP_003829.2
fucose-1-phosphate guanylyltransferase
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FPGT
GST
8790
Wheat germ expression system
25 μg
GFPP
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions