missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ FPGT (Human) Recombinant Protein

Human FPGT full-length ORF ( NP_003829.2, 1 a.a. - 594 a.a.) recombinant protein with GST-tag at N-terminal.

335.00€ - 508.00€

Specifications

Numéro d’adhésion NP_003829.2
Identification génétique (Entrez) 8790
Nom fucose-1-phosphate guanylyltransferase
Méthode de préparation Wheat germ expression system
Test du contrôle qualité 125% SDS-PAGE Stained with Coomassie Blue
View More Specs

Products 2
Product Code Brand Quantité Price Quantity & Availability  
Product Code Brand Quantité Price Quantity & Availability  
16142902
missing translation for 'viewDocuments' Promotion Details
Abnova™
H00008790-P01.10UG
10 μg
335.00€
10µg
View alternative products
missing translation for 'toSaveUpto' 
»
Expédition estimée: 20-06-2024
pour voir le stock disponible 
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
16152902
missing translation for 'viewDocuments' Promotion Details
Abnova™
H00008790-P01.25UG
25 μg
508.00€
25µg
View alternative products
missing translation for 'toSaveUpto' 
»
Expédition estimée: 20-06-2024
pour voir le stock disponible 
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Description

Description

  • Sequence: MAAARDPPEVSLREATQRKLRRFSELRGKLVARGEFWDIVAITAADEKQELAYNQQLSEKLKRKELPLGVQYHVFVDPAGAKIGNGGSTLCALQCLEKLYGDKWNSFTILLIHSGGYSQRLPNASALGKIFTALPLGNPIYQMLELKLAMYIDFPLNMNPGILVTCADDIELYSIGEFEFIRFDKPGFTALAHPSSLTIGTTHGVFVLDPFDDLKHRDLEYRSCHRFLHKPSIEKMYQFNAVCRPGNFCQQDFAGGDIADLKLDSDYVYTDSLFYMDHKSAKMLLAFYEKIGTLSCEIDAYGDFLQALGPGATVEYTRNTSNVIKEESELVEMRQRIFHLLKGTSLNVVVLNNSKFYHIGTTEEYLFYFTSDNSLKSELGLQSITFSIFPDIPECSGKTSCIIQSILDSRCSVAPGSVVEYSRLGPDVSVGENCIISGSYILTKAALPAHSFVCSLSLKMNRCLKYATMAFGVQDNLKKSVKTLSDIKLLQFFGVCFLSCLDVWNLKVTEELFSGNKTCLSLWTARIFPVCSSLSDSVITSLKMLNAVKNKSAFSLNSYKLLSIEEMLIYKDVEDMITYREQIFLEISLKSSLM
Specifications

Specifications

NP_003829.2
fucose-1-phosphate guanylyltransferase
125% SDS-PAGE Stained with Coomassie Blue
GFPP
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
8790
Wheat germ expression system
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FPGT
GST
Videos
SDS
Documents

Documents

Certificates