Learn More
Novus Biologicals™ FPRL2/FPR3 Antibody (7D9B4), Novus Biologicals™
Code produit 30030198 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
0.025 mL
0.1 mL
Conditionnement:
0.03mL
0.10mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30030198 0.025 mL 0.03mL
30029774 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 30030198 Fournisseur Novus Biologicals™ Code fournisseur NBP3271730.025ml
Mouse Monoclonal Antibody
FPRL2/FPR3 Monoclonal antibody specifically detects FPRL2/FPR3 in Human samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spécification
Antigène FPRL2/FPR3
Applications Flow Cytometry, ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 7D9B4
Concentration 1 mg/mL
Conjugué Unconjugated
Dilution Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000
Formule PBS
Alias de gène FMLP-R-II, FMLPY, formyl peptide receptor 3, Formyl peptide receptor-like 2FMLP-related receptor II, FPRH1FPRH2, FPRL2FML2_HUMAN, N-formyl peptide receptor 3, RMLP-R-I
Espèces hôtes Mouse
Immunogène Purified recombinant fragment of human FPRL2/FPR3 expressed in E. Coli.
Méthode de purification Protein G purified
Quantité 0.025 mL
État réglementaire RUO
Disciplines de recherche GPCR, Immunology
Primaire ou secondaire Primary
Identification génétique (Entrez) 2359
Espèces cibles Human
Contenu et stockage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Forme Purified
Isotype IgG1