missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FRZB Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA5117059
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Positive Samples: Mouse skeletal muscle Immunogen sequence: SNGNCRGASS ERCKCKPIRA TQKTYFRNNY NYVIRAKVKE IKTKCHDVTA VVEVKEILKS SLVNIPRDTV NLYTSSGCLC PPLNVNEEYI IMGYEDEERS RLLLVEGSIA EKWKDRLGKK VKRWDMKLRH LGLSKSDSSN SDSTQSQKSG RNSNPRQARN.
The protein encoded by this gene is a secreted protein that is involved in the regulation of bone development. Defects in this gene are a cause of female-specific osteoarthritis (OA) susceptibility.
Spécification
| FRZB | |
| Polyclonal | |
| Unconjugated | |
| FRZB | |
| cb291; Fiz; Fre; FREOS1; frezzled; Fritz; frizzled homolog-related; frizzled protein homolog; frizzled related protein; frizzled-related protein; frizzled-related protein 1; Frp; FRP-3; Frzb; Frzb precursor; FRZB1; FRZB-1; FRZB1sFRP-3; FRZB-PEN; fzb1; fzb-1; FZRB; hFIZ; id:ibd5075; ik:tdsubc_2e7; OS1; secreted frizzled-related protein 3; secreted frizzled-related sequence protein 3; SFRP3; sFRP-3; SFRP3frezzled; SRFP3; xx:tdsubc_2e7 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20378, 2487 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P97401, Q92765 | |
| FRZB | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 166-325 of human FRZB (NP_0014542). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu