missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GABRG3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA141216
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Suggested positive control: rat brain.
GABRG3 is a family of proteins involved in the GABAergic neurotransmission of the mammalian central nervous system. GABRG3 is a member of the GABA-A receptor gene family of heteromeric pentameric ligand-gated ion channels through which GABA, the major inhibitory neurotransmitter in the mammalian brain, acts. GABA-A receptors are the site of action of a number of important pharmacologic agents including barbiturates, benzodiazepines, and ethanol (summary by Whiting et al., 1999). For additional general information about the GABA-A receptor gene family, see GABRA1 (MIM 137160).
Spécification
| GABRG3 | |
| Polyclonal | |
| Unconjugated | |
| GABRG3 | |
| B230362M20Rik; CAE2; ECA2; GABA(A) receptor subunit gamma-3; GABA(G) receptor; GABA(G) receptor, gamma 3; GABA-alpha receptor gamma-3 subunit; GABRG3; Gabrg-3; gamma-aminobutyric acid; gamma-aminobutyric acid (GABA) A receptor, gamma 3; gamma-aminobutyric acid (GABA) A receptor, subunit gamma 3; gamma-aminobutyric acid (GABA-A) receptor, subunit gamma 3; gamma-aminobutyric acid A receptor, gamma 3; gamma-aminobutyric acid receptor subunit gamma-3; gamma-aminobutyric acid type A receptor gamma3 subunit; GEFSP3 | |
| Rabbit | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| Conc. Not Determined | |
| PBS with 0.05% sodium azide | |
| P27681, P28473, Q99928 | |
| GABRG3 | |
| A synthetic peptide corresponding to amino acids somewhere between 10-60 of human GABAA receptor. | |
| RUO | |
| 14407, 2567, 79211 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu