missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAPDH Antibody (13H12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 29 publications
Marque: Novus Biologicals NBP2-27103
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
GAPDH Monoclonal specifically detects GAPDH in Human, Mouse, Rat, Drosophila, Monkey, Primate, Sheep samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Spécification
| GAPDH | |
| Monoclonal | |
| 1.0 mg/ml | |
| Western Blot 0.25 - 1 ug/ml, Simple Western 1:25, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 1:10, Immunohistochemistry-Paraffin 5 ug/ml | |
| aging-associated gene 9 protein, EC 1.2.1, EC 1.2.1.12, EC 2.6.99.-, G3PD, GAPD, glyceraldehyde 3-phosphate dehydrogenase, glyceraldehyde-3-phosphate dehydrogenase, MGC88685, Peptidyl-cysteine S-nitrosylase GAPDH | |
| Mouse | |
| 36 kDa | |
| 0.1 mg | |
| Alzheimers Research, Apoptosis, Autophagy, Cancer, DNA Repair, Lipid and Metabolism, Loading Controls, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Nuclear Receptors Coactivators and Corepressors, Signal Transduction, Translation Control | |
| 2597 | |
| Human, Mouse, Rat, Drosophila, Primate, Sheep | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 13H12 | |
| Unconjugated | |
| P04406 | |
| GAPDH | |
| Amino acids between 275 and 325 of glyceraldehyde 3-phosphate dehydrogenase protein were used as the immunogen for this GAPDH antibody. | |
| Protein G purified | |
| RUO | |
| Primary | |
| Reacts with Human, Primates/Monkey, Mouse and Drosophila. Based upon 91% sequence similarity with immunogen, this antibody is predicted to react with Guinea Pig, Sheep, Squirrel, Porcine/Pig, Ferret, Canine/Dog/Cat, Bovine, Reptile / Rattlesnake and several other species. Immunogen shows 82% similarity to Xenopus and Zebrafish. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu