missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCN1L1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP1-83382
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
GCN1L1 Polyclonal specifically detects GCN1L1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Spécification
| GCN1L1 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/ml, Immunohistochemistry 1:20 - 1:50, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| GCN1, GCN1 (general control of amino-acid synthesis 1, yeast)-like 1, GCN1 general control of amino-acid synthesis 1-like 1 (yeast), GCN1L, GCN1-like protein 1, HsGCN1, KIAA0219hsGCN1, peroxisome proliferator activated receptor interacting complex protein, PRIC295, translational activator GCN1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 10985 | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| GCN1L1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ALKEKLGTPDEQLEMANCQAVILSVEDDTGHRIIIEDLLEATRSPEVGMRQAAAIILNIYCSRSKADYTSHLRSLVSGLIRLFNDSSPVVLEESWDALNAITKKLDAGNQLALIEELHKEIRLIGNESKGEHVPGFC | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu