missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Gelsolin Recombinant Rabbit Monoclonal Antibody (HL1931)

Rabbit Recombinant Monoclonal Antibody
Marque: Invitrogen™ MA547226
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
The protein encoded by this gene binds to the plus ends of actin monomers and filaments to prevent monomer exchange. The encoded calcium-regulated protein functions in both assembly and disassembly of actin filaments. Defects in this gene are a cause of familial amyloidosis Finnish type (FAF). Multiple transcript variants encoding several different isoforms have been found for this gene.
Spécification
| Gelsolin | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| P06396 | |
| GSN | |
| Recombinant fragment of human Gelsolin. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| HL1931 | |
| Unconjugated | |
| GSN | |
| Actin-depolymerizing factor; ADF; ADFBrevin; AGEL; Brevin; DKFZp313L0718; gelsolin; gelsolin (amyloidosis, Finnish type); Gsb; Gsn; hypothetical protein LOC535077; plasma gelsolin precursor; RP11-477J21.1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 2934 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu