missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GFP Antibody (9F9.F9), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
Marque: Novus Biologicals NB110-40670
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
GFP Monoclonal specifically detects GFP in Non-species specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.
Spécification
| GFP | |
| Monoclonal | |
| 1.0mg/mL | |
| Western Blot 1:2000-1:10000, ELISA 1:50000-1:200000, Immunohistochemistry 1:1000-1:5000, Immunohistochemistry-Paraffin 1:10-1:500, Knockdown Validated | |
| GFP | |
| Recombinant Green Fluorescent Protein (GFP) fusion protein corresponding to the full length amino acid sequence (246 aa) derived from the jellyfish Aequorea victoria. | |
| Protein A purified | |
| RUO | |
| Reactivity is observed against recombinant Green Fluorescent Protein from Aequorea victoria by both Western blot and ELISA. No reaction is seen against RFP. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown | |
| 9F9.F9 | |
| Biotin | |
| Enhanced Green Fluorescent Protein, Green Fluorescent Protein, green fluorescent protein (gfp) | |
| Mouse | |
| 27 kDa | |
| 0.5 mg | |
| Primary | |
| Jellyfish | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu