missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLS2 Antibody (CL7721) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP2-88908PCP
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
GLS2 Monoclonal antibody specifically detects GLS2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Spécification
| GLS2 | |
| Monoclonal | |
| PerCP | |
| EC 3.5.1.2, GAbreast cell glutaminase, GLSglutaminase GA, glutaminase 2 (liver, mitochondrial), glutaminase I, glutaminase liver isoform, mitochondrial, hLGA, LGA, L-glutaminase, L-glutamine amidohydrolase, MGC71567, phosphate-activated glutaminase, phosphate-dependent glutaminase, truncated glutaminase 2 | |
| This antibody was developed using a synthetic peptide derived from Q9UI32, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
| 27165 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL7721 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu