missing translation for 'onlineSavingsMsg'
Learn More

Glucose 6 phosphate isomerase Antibody (CPTC-GPI-1), PE, Novus Biologicals™

Code produit 18615321 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
0.1 mL
Conditionnement:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18615321 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 18615321 Fournisseur Novus Biologicals Code fournisseur NBP279845PE

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Mouse Monoclonal Antibody

Glucose 6 phosphate isomerase Monoclonal antibody specifically detects Glucose 6 phosphate isomerase in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array
TRUSTED_SUSTAINABILITY

Spécification

Antigène Glucose 6 phosphate isomerase
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone CPTC-GPI-1
Conjugué PE
Formule PBS
Alias de gène AMFGNPI, Autocrine motility factor, DKFZp686C13233, EC 5.3.1.9, glucose phosphate isomerase, glucose-6-phosphate isomerase, hexose monophosphate isomerase, hexosephosphate isomerase, neuroleukin, NLKSA36, oxoisomerase, PGI, PHI, Phosphoglucose isomerase, phosphohexomutase, Phosphohexose isomerase, phosphosaccharomutase, SA-36, Sperm antigen 36, sperm antigen-36
Espèces hôtes Mouse
Immunogène Recombinant human full-length Glucose 6 phosphate isomerase protein (Uniprot: P06744)
Méthode de purification Protein A or G purified
Quantité 0.1 mL
État réglementaire RUO
Disciplines de recherche Cytokine Research, Phospho Specific
Primaire ou secondaire Primary
Identification génétique (Entrez) 2821
Espèces cibles Human
Contenu et stockage Store at 4°C in the dark.
Forme Purified
Isotype IgG2a κ
Afficher plus Afficher moins
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.