missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
Glut1 Polyclonal antibody specifically detects Glut1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
| Antigène | Glut1 |
| Applications | Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugué | Unconjugated |
| Dilution | Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/ Immunofluorescence 0.25 - 2 μg/mL, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Formule | PBS (pH 7.2), 40% Glycerol |
| Alias de gène | Choreoathetosis/Spasticity, Episodic (Paroxysmal Choreoathetosis), CSE, DYT17, DYT18, DYT9, EIG12, Glucose transporter type 1, erythrocyte/brain, GLUT-1, GLUT1DS, HepG2 glucose transporter, HTLVR, Human T-Cell Leukemia Virus (I and II) Receptor, MGC141895, MGC141896, PED, solute carrier family 2 (facilitated glucose transporter), member 1, Solute Carrier Family 2 Member 1, solute carrier family 2, facilitated glucose transporter member 1 |
| Espèces hôtes | Rabbit |
| Immunogène | This Glut1 antibody was developed against a recombinant protein corresponding to amino acids: GRTFDEIASGFRQGGASQSDKTPEELFHP |
| Méthode de purification | Immunogen affinity purified |
| Mostrar más |
Título del producto
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?