missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glut4 Antibody (925932) [FITC], Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals FAB86541F
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Glut4 Monoclonal antibody specifically detects Glut4 in Human samples. It is validated for Flow Cytometry, CyTOF
Spécification
| Glut4 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 925932 | |
| Flow Cytometry, CyTOF-ready | |
| GLUT-4, GLUT4Glucose transporter type 4, insulin-responsive, insulin-responsive glucose transporter type 4, solute carrier family 2 (facilitated glucose transporter), member 4, solute carrier family 2, facilitated glucose transporter member 4 | |
| Mouse myeloma cell line NS0-derived recombinant human Glut4, Met1-Asp509, Accession # P14672 | |
| 0.1 mL | |
| Cellular Markers, Core ESC Like Genes, Diabetes Research, Hematopoietic Stem Cell Markers, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neuroendocrinology, Neuronal Cell Markers, Neuroscience, Stem Cell Markers | |
| 6517 | |
| Store at 4°C in the dark. | |
| IgG2b |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur