missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GM2 Ganglioside Antibody (KM966) - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Marque: Novus Biologicals NBP3-43552
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
GM2 Ganglioside Monoclonal antibody specifically detects GM2 Ganglioside in Human samples. It is validated for Flow Cytometry,ELISA,Immunocytochemistry/ Immunofluorescence
Spécification
| GM2 Ganglioside | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence | |
| GM2 (ganglioside), Tay-Sachs ganglioside | |
| This chimeric antibody was generated by cloning the variable heavy and light chain sequences of murine parent antibody KM696 (IgM) and expressing it as a mouse/human IgG1. The parent antibody was raised against human GM2. | |
| 0.2 mg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
| KM966 | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu