Learn More
GM2 Ganglioside Antibody (L55-81), PerCP, Novus Biologicals™
Rabbit Monoclonal Antibody
Marque: Novus Biologicals NBP2-81278PCP
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
GM2 Ganglioside Monoclonal antibody specifically detects GM2 Ganglioside in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Functional Assay, Cell depletion
Spécification
| GM2 Ganglioside | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4C in the dark. | |
| IgG κ |
| ELISA, Immunohistochemistry, Immunocytochemistry, Functional Assay, Depletion Assay | |
| L55-81 | |
| ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Functional, Cell depletion | |
| GM2 (ganglioside), Tay-Sachs ganglioside | |
| L55-81 was prepared by isolating EBV-immortalized peripheral blood B lymphocytes from a human donor with melanoma. | |
| 0.1 mL | |
| Cancer | |
| Human | |
| Purified |