Learn More
GnRH1 Monoclonal Antibody (LHRH13-372.14), Invitrogen™
Mouse Monoclonal Antibody
Marque:  Invitrogen MA5-46175
Informations supplémentaires : Poids : 0.01000kg
 Afficher plus de versions de ce produit
Code produit. 17890176
  • 369.00€ / 1mg
Expédition estimée: 26-10-2023
pour voir le stock disponible 
Description
Description
MA1-10815 detects Gonadotropin Releasing Hormone from human samples. MA1-10815 has been successfully used in ELISA and immunohistochemistry applications. The MA1-10815 immunogen is purified human gonadotropin releasing hormone.
The protein encoded by this gene is secreted and then cleaved to form the 10 aa luteinizing hormone-releasing hormone (LHRH), also known as gonadoliberin-1, and prolactin release-inhibiting factor, also known as GnRH-associated peptide 1. LHRH stimulates the release of luteinizing and follicle stimulating hormones, which are important for reproduction. Two transcript variants encoding the same protein have been found for this gene.
Spécification
Spécification
GnRH1
Monoclonal
1.74 mg/mL
GNRH1
GNRH1
Purified human gonadotropin releasing hormone.
1 mg
Primary
Human
Antibody
IgG1
ELISA, Immunohistochemistry
LHRH13-372.14
Unconjugated
P01148
Mouse
Purified
RUO
2796
-20° C, Avoid Freeze/Thaw Cycles