Learn More
Invitrogen™ GnRH1 Monoclonal Antibody (LHRH13-372.14), Invitrogen™
Mouse Monoclonal Antibody
Marque:  Invitrogen™ MA5-46175
Code nomenclature Nacres: NA.46
Code produit. 17890176
  • 1052.00€ / 1mg
Description
MA1-10815 detects Gonadotropin Releasing Hormone from human samples. MA1-10815 has been successfully used in ELISA and immunohistochemistry applications. The MA1-10815 immunogen is purified human gonadotropin releasing hormone.
The Luteinizing Hormone Releasing Hormone (LH-RH) is produced in the hypothalamus and influences the release of Luteinizing Hormone in the pituitary gland. LH-RH is also known as Gonadotropin-releasing hormone (Gn-RH).
TRUSTED_SUSTAINABILITY
Spécification
Spécification
GnRH1
Monoclonal
1.40 mg/mL
PBS with no preservative; pH 7.4
P01148
GNRH1
Purified human gonadotropin releasing hormone.
1 mg
Primary
Human
Antibody
IgG1
ELISA, Immunohistochemistry
LHRH13-372.14
Unconjugated
GNRH1
Gnrh; Gnrh1; Gnrh2; Gnrha; GnRH-associated peptide 1; GnRH-associated peptide I; GnRH-I; Gonadoliberin I; Gonadoliberin-1; Gonadorelin; gonadotropin releasing hormone 1; gonadotropin releasing hormone 2; gonadotropin-releasing hormone 1 (luteinizing-releasing hormone); gonadotropin-releasing hormone 1 variant A; gonadotropin-releasing hormone 1 variant B; Gonadotropin-releasing hormone I; GRH; hpg; hypogonadal; leuteinizing-releasing hormone; LHRH; LH-RH I; Lhrh1; LNRH; Luliberin I; luteinizing hormone releasing hormone; luteinizing hormone-releasing hormone; luteinizing hormone-releasing hormone I; Lutropin; Progonadoliberin I; progonadoliberin-1; prolactin release-inhibiting factor; Prolactin release-inhibiting factor 1; Prolactin release-inhibiting factor I; Rgnrhg1
Mouse
Protein A
RUO
2796
-20°C, Avoid Freeze/Thaw Cycles