missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Guinea pig IgG F(ab')2 fragment, Texas Red, Polyclonal, Secondary Antibody, Abnova™
Marque: Abnova PAB10619.2mg
Informations supplémentaires : Poids : 0.00010kg
Spécification
Goat anti-Guinea pig IgG F(ab')2 fragment | |
Polyclonal | |
Goat anti-guinea pig IgG recognizes Guinea pig IgG F(ab')2 fragment. This secondary antibody was purified using antigen affinity chromatography. The antibody is conjugated with Texas Red. | |
Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
Guinea pig IgG F(ab')2 fragment | |
2 mg | |
Guinea Pig | |
Lyophilized |
Flow Cytometry, Immunofluorescence | |
Texas Red | |
The optimal working dilution should be determined by the end user. | |
Goat | |
This product was prepared from monospecific antiserum by immunoaffinity chromatography, followed by solid phase adsorption(s) to remove any unwanted reactivities. | |
Secondary | |
Store at 4°C on dry atmosphere. After reconstitution with 1 mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
For Research Use Only