missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Golgin-97 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA583719
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Immunogen sequence: TGAESKITAL EQKEQELQAL IQQLSIDLQK VTAETQEKED VITHLQEKVA SLEKRLEQNL SGEEHLQELL K Highest antigen sequence indentity to the following orthologs: Mouse - 85%, Rat - 80%.
The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. The Golgins are a family of proteins, of which the protein encoded by this gene is a member, that are localized to the Golgi. This encoded protein is associated with Sjogren's syndrome.
Spécification
| Golgin-97 | |
| Polyclonal | |
| Unconjugated | |
| GOLGA1 | |
| 0710001G09Rik; 2210418B03Rik; AW107649; gap junction protein, alpha 4, 37kD; Golga1; golgi autoantigen, golgin subfamily a, 1; Golgi97; golgin A1; golgin subfamily A member 1; golgin-97 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2800 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q92805 | |
| GOLGA1 | |
| Recombinant protein corresponding to Human GOLGA1. Recombinant protein control fragment (Product #RP-102964). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu