missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR61 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA533744
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Elephant, Panda, Pig (100%) Marmoset, Bovine (94%) Bat (88%).
G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. GPCR GPR61 is a member of this family (subfamily Orphan-A). It has been reported to be expressed in several regions of brain, including caudate, putamen, and thalamus.
Spécification
| GPR61 | |
| Polyclonal | |
| Unconjugated | |
| GPR61 | |
| BALGR; biogenic amine receptor-like GPCR; Biogenic amine receptor-like G-protein coupled receptor; G protein-coupled receptor 61; GPCR3; GPR61; G-protein coupled receptor 61; probable G-protein coupled receptor 61 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100514405, 83873 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9BZJ8 | |
| GPR61 | |
| Synthetic 16 amino acid peptide from N-terminal extracellular domain of human GPR61. | |
| 50 μg | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu