missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPRC6A Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA5102095
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Antibody detects endogenous levels of total GPRC6A.
GPRC6A is an Orphan-U GPCR with an unknown ligand. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more thana thousandmembers currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This turn-off activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates. Twoblockbuster drugs which have been developed as GPCR-targetedpharmaceuticals are Zyprexa (Eli Lilly) and Claritin (Schering-Plough) which have multi-billion dollar shares of the mental health and allergy markets, respectively.
Spécification
| GPRC6A | |
| Polyclonal | |
| Unconjugated | |
| GPRC6A | |
| bA86F4.3; G protein-coupled receptor class C group 6 member A; G protein-coupled receptor, class C, group 6, member A; G protein-coupled receptor, family C, group 6, member A; GPCR; Gprc6a; G-protein coupled receptor family C group 6 member A; G-protein coupled receptor GPCR33; hGPCR33; hGPRC6A; predicted with SOSUI analysis; seven transmembrane helix receptor | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 222545 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5T6X5 | |
| GPRC6A | |
| A synthesized peptide derived from human GPRC6A(Accession Q5T6X5), corresponding to amino acid residues P504-T554. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu