missing translation for 'onlineSavingsMsg'
Learn More

GPRC6A Rabbit anti-Human, Polyclonal, Novus Biologicals™

Code produit 18391951 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
0.1 mL
Conditionnement:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18391951 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 18391951 Fournisseur Novus Biologicals Code fournisseur NBP2822430.1ML

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Polyclonal Antibody

GPRC6A Polyclonal specifically detects GPRC6A in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Spécification

Antigène GPRC6A
Applications Western Blot
Classification Polyclonal
Conjugué Unconjugated
Formule PBS, 2% sucrose
Alias de gène bA86F4.3, G protein-coupled receptor, family C, group 6, member A, GPCR, G-protein coupled receptor family C group 6 member A, G-protein coupled receptor GPCR33, hGPCR33, hGPRC6A, predicted with SOSUI analysis, seven transmembrane helix receptor
Espèces hôtes Rabbit
Immunogène The immunogen is a synthetic peptide directed towards the N-terminal region of Human GPRC6A
Méthode de purification Immunogen affinity purified
Quantité 0.1 mL
État réglementaire RUO
Disciplines de recherche GPCR
Primaire ou secondaire Primary
Identification génétique (Entrez) 222545
Espèces cibles Human
Contenu et stockage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Forme Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.