missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Gram Negative Endotoxin Monoclonal Antibody (GNE13-305.6), Invitrogen™
Mouse Monoclonal Antibody
424.00€ - 1052.00€
Spécification
| Antigène | Gram Negative Endotoxin |
|---|---|
| Clone | GNE13-305.6 |
| Concentration | 1.8 mg/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
|
17800296
|
Invitrogen™
MA5-46002 |
100 μg |
424.00€
100µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
|
11518952
|
Invitrogen™
MA110685 |
500 μg |
570.00€
500µg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
|
17890216
|
Invitrogen™
MA5-46079 |
1 mg |
1052.00€
1mg |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
MA1-10685 detects Endotoxin from E. coli, Sh. sonnei, and S. typhimurium samples. MA1-10685 has been successfully used in ELISA applications. The MA1-10685 immunogen is E. coli serotypes: O:111, B4 J5.
Endotoxins are part of the outer membrane of the cell wall of gram-negative bacteria. Endotoxins are invariably associated with gram-negative bacteria whether the organisms are pathogens or not. Although the term "e;endotoxin"e; is occasionally used to refer to any cell-associated bacterial toxin, it is properly reserved to refer to the lipopolysaccharide complex associated with the outer membrane of gram-negative bacteria such as E. coli, Salmonella, Shigella, Pseudomonas, Neisseria, Haemophilus, and other leading pathogens. The biological activity of endotoxin is associated with the lipopolysaccharide (LPS). Toxicity is associated with the lipid component (Lipid A) and immunogenicity is associated with the polysaccharide components. The cell wall antigens (O antigens) of gram-negative bacteria are components of LPS. LPS elicits a variety of inflammatory responses in an animal. Because it activates complement by the alternative (properdin) pathway, it is often part of the pathology of gram-negative bacterial infections.Spécification
| Gram Negative Endotoxin | |
| 1.8 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.2-7.4 | |
| IgG2a | |
| Protein A/G | |
| Antibody |
| GNE13-305.6 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Bacteria | |
| E. coli serotypes: O:111, B4 J5. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit