missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Granulocytes Monoclonal Antibody (6D10)

Mouse Monoclonal Antibody
Marque: Invitrogen™ MA528457
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Mouse anti Pig Granulocytes antibody, clone 6D10 recognizes a approximately 60 kDa antigen on porcine granulocytes of the neutrophil lineage, acting as a reliable tool for their analysis and isolation, without contamination from other cells.
Granulocytes are a category of white blood cells characterized by the presence of granules in their cytoplasm. They are also called polymorphonuclear leukocytes (PMN or PML) because of the varying shapes of the nucleus, which is usually lobed into three segments. In common parlance, the term polymorphonuclear leukocyte often refers specifically to neutrophil granulocytes, the most abundant of the granulocytes. Granulocytes or PMN are released from the bone marrow by the regulatory complement proteins.
Spécification
| Granulocytes | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| Mouse | |
| Protein G | |
| RUO | |
| Pig | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunoprecipitation, Western Blot | |
| 6D10 | |
| Unconjugated | |
| C17orf33; colony stimulating factor 3; colony stimulating factor 3 (granulocyte); CSF3; CSF3OS; filgrastim; GCSF; G-CSF; Granulocyte colony-stimulating factor; Granulocyte Marker; granulocyte-colony stimulating factor; lenograstim; Pluripoietin; PML; PMN | |
| Porcine bone marrow hematopoietic cells (BMHC). | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu