missing translation for 'onlineSavingsMsg'
Learn More

GREB1 Antibody (CL13859) [Biotin], Novus Biologicals™

Code produit 30524626 Tous les produits Bio Techne Produits
Change view
Click to view available options
Quantité:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité unitSize
30524626 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30524626 Supplier Novus Biologicals Supplier No. NBP343839B

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

GREB1 Monoclonal antibody specifically detects GREB1 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigène GREB1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL13859
Conjugué Biotin
Formule PBS
Alias de gène gene regulated by estrogen in breast cancer protein, growth regulation by estrogen in breast cancer 1, KIAA0575Gene regulated in breast cancer 1 protein, protein GREB1
Espèces hôtes Mouse
Immunogène This antibody was generated using a recombinant protein sequence of Q4ZG55, with the exact immunogen sequence remaining proprietary.
Méthode de purification Protein A purified
Quantité 0.1 mL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 9687
Espèces cibles Human
Contenu et stockage Store at 4°C in the dark.
Forme Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.