missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRF2 Antibody (OTI2F5), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Marque: Novus Biologicals NBP2-70846PE
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
GRF2 Monoclonal antibody specifically detects GRF2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Spécification
| GRF2 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| OTI2F5 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| C3GGRF2guanine nucleotide-releasing factor 2 (specific for crk proto-oncogene), CRK SH3-binding GNRP, DKFZp781P1719, Guanine nucleotide-releasing factor 2, Protein C3G, Rap guanine nucleotide exchange factor (GEF) 1, rap guanine nucleotide exchange factor 1 | |
| Full length human recombinant protein of human RAPGEF1(NP_941372) produced in HEK293T cell. | |
| 0.1 mL | |
| Signal Transduction | |
| 2889 | |
| Store at 4C in the dark. | |
| IgG1 |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu