missing translation for 'onlineSavingsMsg'
Learn More

GSC2 Antibody, Novus Biologicals™

Product Code. p-200070836 Shop All Bio Techne Products
Change view
Click to view available options
Quantité:
25 μL
Unit Size:
25µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité unitSize
18414192 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414192 Supplier Novus Biologicals Supplier No. NBP19197025ul

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GSC2 Polyclonal specifically detects GSC2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigène GSC2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugué Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200
Formule PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Alias de gène goosecoid homeobox 2, goosecoid-like, GSC-2, GSC-L, GSCLhomeobox protein goosecoid-2, Homeobox protein goosecoid-like
Symboles de gène(s) GSC2
Espèces hôtes Rabbit
Immunogène This antibody was developed against Recombinant Protein corresponding to amino acids:PFSIEHILSSLPERSLPARAACPPQPAGRQSPAKPEEP
Méthode de purification Affinity Purified
Quantité 25 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 2928
Spécificité du test Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Espèces cibles Human
Contenu et stockage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.