Learn More
Invitrogen™ HA Tag Monoclonal Antibody (2-2.2.14)
Mouse Monoclonal Antibody
Marque: Invitrogen™
880.10 EUR valable jusqu'au 2025-12-19
MEILLEUR PRIX promo! Utilisez le code promo "24092" pour bénéficier de cette offre.
Description
26183 is specific for the HA peptide YPYDVPDYA. The antibody has been used successfully in Western blot and immunoprecipitation applications. HA Synthetic Peptide (Product # 26184) is available for use in neutralization and control experiments and to competitively elute HA-tagged fusion proteins from immobilized anti-HA affinity resin.
HA Tag (hemagglutinin tag) is an epitope tag that provides a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. HA (hemagglutinin) is a surface glycoprotein derived from influenza hemagglutinin, and is commonly expressed as a tag in conjunction with proteins to aid in identification and purification of the tagged protein for functional analysis. HA tag antibodies can be used to facilitate protein detection or co-immunopreciptation of tagged proteins.
Spécification
| HA Tag | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide; pH 7.4 | |
| Mouse | |
| Protein G | |
| RUO | |
| Tag | |
| Antibody | |
| IgG1 |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 2-2.2.14 | |
| Unconjugated | |
| HA; HA Epitope Tag; HA Tag; HA-tag; hemagglutinin | |
| HA peptide YPYDVPDYA derivitized to ovalbumin. | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| Tag |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.