Learn More
HAI-2/SPINT2 Antibody (1), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Marque: Novus Biologicals (Bio-Techne) NBP2-90373CL1
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
HAI-2/SPINT2 Monoclonal antibody specifically detects HAI-2/SPINT2 in Mouse samples. It is validated for ELISA
Spécification
| HAI-2/SPINT2 | |
| Monoclonal | |
| CoraFluor 1 | |
| HAI-2DIAR3, HAI2FLJ45571, Hepatocyte growth factor activator inhibitor type 2, KOP, kunitz-type protease inhibitor 2, PB, Placental bikunin, serine peptidase inhibitor, Kunitz type, 2, serine protease inhibitor, Kunitz type, 2 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse HAI-2/SPINT2 extracellular domain (Accession#: NP_001076017.1; Met 1-Lys 140). | |
| 0.1 mL | |
| Cancer | |
| 10653 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA | |
| 1 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |