Learn More
Herpes Simplex Virus 1 (HSV1) Mouse anti-Virus, Clone: HSVI/2095, Novus Biologicals™
Mouse Monoclonal Antibody
260.00€
Spécification
| Antigène | Herpes Simplex Virus 1 (HSV1) |
|---|---|
| Clone | HSVI/2095 |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Immunohistochemistry (Paraffin) |
Description
Herpes Simplex Virus 1 (HSV1) Monoclonal specifically detects Herpes Simplex Virus 1 (HSV1) in Virus samples. It is validated for Immunohistochemistry-Paraffin.Spécification
| Herpes Simplex Virus 1 (HSV1) | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Virus | |
| Herpes simplex virus 1, Herpes Simplex Virus 1 and 2, Herpes simplex virus 2, Herpes simplex virus type 1, Herpes simplex virus type 2, HSV 1, HSV 2 | |
| IgG1 κ | |
| Protein A or G purified |
| HSVI/2095 | |
| Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| Baculovirus-expressed HSV DNA polymerase (POL) and POL/UL42 complex | |
| Primary | |
| Store at 4°C. |