missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HEXIM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marque: Invitrogen™ PA552629
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Immunogen sequence: AKSDDTSDDD FMEEGGEEDG GSDGMGGDGS EFLQRDFSET YERYHTESLQ NMSKQELIKE YLELEKCLSR MEDENNRLRL ESKRLGGDDA RVRELELELD RLRAENLQLL TENELHRQQE RA Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.
The protein encoded by this gene is a member of the RecQ DNA helicase family. DNA helicases are enzymes involved in various types of DNA repair, including mismatch repair, nucleotide excision repair and direct repair. Some members of this family are associated with genetic disorders with predisposition to malignancy and chromosomal instability. The biological function of this helicase has not yet been determined. Two alternatively spliced transcripts, which encode the same isoform but differ in their 5' and 3' UTRs, have been described.
Spécification
| HEXIM1 | |
| Polyclonal | |
| Unconjugated | |
| HEXIM1 | |
| 7330426E13Rik; Cardiac lineage protein 1; CLP1; Clp-1; EDG1; Estrogen down-regulated gene 1 protein; FLJ13562; hexamethylene bisacetamide inducible 1; hexamethylene bis-acetamide inducible 1; hexamethylene bisacetamide-inducible protein; hexamethylene bis-acetamide-inducible protein 1; hexamethylene-bis-acetamide-inducible transcript 1; hexamthylene bis-acetamide inducible 1; HEXIM1; HGNC:24953; HIS1; HMBA-inducible; MAQ1; menage a quatre 1; menage a quatre protein 1; Protein HEXIM1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10614 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O94992 | |
| HEXIM1 | |
| Recombinant protein corresponding to Human HEXIM1. Recombinant protein control fragment (Product #RP-89427). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu