missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ HGF Antibody (016) [Allophycocyanin], Novus Biologicals™
Rabbit Monoclonal Antibody
Marque: Novus Biologicals™ NBP3-26881APC
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
HGF Monoclonal antibody specifically detects HGF in Human samples. It is validated for ELISA
Spécification
| HGF | |
| Monoclonal | |
| APC | |
| deafness, autosomal recessive 39, DFNB39, EC 3.4.21, EC 3.4.21.7, fibroblast-derived tumor cytotoxic factor, F-TCF, hepatocyte growth factor (hepapoietin A; scatter factor), Hepatopoeitin-A, HGFB, HPTAhepatocyte growth factor, lung fibroblast-derived mitogen, Scatter factor, SFhepatopoeitin-A | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human HGF (RefSeq#: NP_000592.3; Met1-Ser728). | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 3082 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 016 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu