missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HH3 Monoclonal Antibody (4E9B11)

Mouse Monoclonal Antibody
Marque: Invitrogen™ MA531758
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
MA5-31758 has been tested in indirect ELISA.
Histones are relatively small proteins that contain a high proportion of positively charged amino acids, e. g., arginine and lysine. This positive charge allows histones to bind tightly to the negatively charged DNA. There are five basic types of histone molecules called H1, H2A, H2B, H3, and H4. Histones are abundantly present in virtually all eukaryotic cells.
Spécification
| HH3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q16695 | |
| H3-4, Hist3 | |
| Synthesized peptide of human HH3 (AA: ARTKQTAR(AcK)STG-C) (aa 22-36). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot | |
| 4E9B11 | |
| Unconjugated | |
| H3-4 | |
| H3 histone family, member T; H3.4; H3/g; H3/t; H3FT; H3t; HIST3H3; histone 3, H3; histone cluster 3, H3; histone H3.1t | |
| Mouse | |
| Protein G | |
| RUO | |
| 15268, 8290 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu