missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIST3H3 Antibody (2128A), mFluor Violet 610 SE, Novus Biologicals™
Tous les produits Bio Techne ProduitsDescription
HIST3H3 Monoclonal antibody specifically detects HIST3H3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry
Spécification
Spécification
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry |
| Conjugué | mFluor Violet 610 SE |
| Contenu et stockage | Store at 4C in the dark. |
Nom du produit
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?